Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- EDEM2
- Entrez Gene ID
-
55741
- Gene Name
- ER degradation enhancing alpha-mannosidase like protein 2
- Gene Aliases
-
C20orf31, C20orf49, bA4204.1
- Chromosome Location
-
Chr.20: 35115357 - 35147358 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK293583.1 |
- |
5 - 6 |
677 |
64 |
View Full Product Details