Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EDEM2
Entrez Gene ID
55741
Gene Name
ER degradation enhancing alpha-mannosidase like protein 2
Gene Aliases
C20orf31, C20orf49, bA4204.1
Chromosome Location
Chr.20: 35115357 - 35147358 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145025.1 NP_001138497.1 7 - 8 953 72
NM_018217.2 NP_060687.2 8 - 9 1064 72
NR_026728.1 7 - 8 1280 72
GenBank mRNA AK001645.1 - 8 - 9 1046 72
AK023931.1 - 8 - 9 1034 72
AK096016.1 - 6 - 7 974 72
AK293583.1 - 7 - 8 935 72
AK298794.1 - 7 - 8 1022 72
AK300212.1 - 7 - 8 908 72
AK303816.1 - 6 - 7 706 72
AY007088.1 - 5 - 6 632 72
AY358580.1 - 7 - 8 914 72
BC001371.2 - 8 - 9 1031 72
BC016184.1 - 8 - 9 1005 72
CR457266.1 - 8 - 9 964 72

View Full Product Details