The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014547.4 | NP_055362.1 | 5 - 6 | 780 | 136 |
XM_017022087.1 | XP_016877576.1 | 6 - 7 | 811 | 136 | |
XM_017022088.1 | XP_016877577.1 | 5 - 6 | 652 | 136 | |
GenBank mRNA | AF177171.1 | - | 5 - 6 | 742 | 136 |
AF237631.1 | - | 4 - 5 | 562 | 136 | |
AK310344.1 | - | 3 - 4 | 459 | 136 | |
AK312569.1 | - | 5 - 6 | 777 | 136 | |
BC020542.2 | - | 5 - 6 | 716 | 136 | |
BX647277.1 | - | 5 - 6 | 742 | 136 |