Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ITGA3
Entrez Gene ID
3675
Gene Name
integrin subunit alpha 3
Gene Aliases
CD49C, FRP-2, GAP-B3, GAPB3, ILNEB, MSK18, VCA-2, VL3A, VLA3a
Chromosome Location
Chr.17: 50055968 - 50090485 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002204.3 NP_002195.1 17 - 18 2696 72
XM_005257308.2 XP_005257365.1 15 - 16 2194 72
GenBank mRNA AB209658.1 - 15 - 16 3145 72
AK289961.1 - 17 - 18 2322 72
AK303384.1 - 18 - 19 2250 72
BC136636.1 - 17 - 18 2296 72
BC144328.1 - 17 - 18 2296 72
BC150190.1 - 17 - 18 2539 72
D01038.1 - 17 - 18 2128 72
M59911.1 - 17 - 18 2297 72

View Full Product Details