The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002204.3 | NP_002195.1 | 19 - 20 | 2869 | 111 |
XM_005257308.2 | XP_005257365.1 | 17 - 18 | 2367 | 111 | |
GenBank mRNA | AB209658.1 | - | 17 - 18 | 3318 | 111 |
AK289961.1 | - | 19 - 20 | 2495 | 111 | |
AK303384.1 | - | 20 - 21 | 2423 | 111 | |
BC136636.1 | - | 19 - 20 | 2469 | 111 | |
BC144328.1 | - | 19 - 20 | 2469 | 111 | |
BC150190.1 | - | 19 - 20 | 2712 | 111 | |
D01038.1 | - | 19 - 20 | 2301 | 111 | |
M59911.1 | - | 19 - 20 | 2470 | 111 |