Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TEX10
Entrez Gene ID
54881
Gene Name
testis expressed 10
Gene Aliases
Ipi1, bA208F1.2
Chromosome Location
Chr.9: 100302075 - 100352977 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161584.1 NP_001155056.1 9 - 10 2134 135
NM_017746.3 NP_060216.2 9 - 10 2190 135
XM_011518798.1 XP_011517100.1 7 - 8 1777 135
XM_017014848.1 XP_016870337.1 9 - 10 1679 135
GenBank mRNA AB060968.1 - 9 - 10 2106 135
AJ420436.1 - 4 - 5 615 135
AK000294.1 - 9 - 10 2106 135
AK298916.1 - 11 - 12 1713 135
AK302619.1 - 9 - 10 2137 135
AY598337.1 - 8 - 9 1975 135
BC030652.2 - 9 - 10 2026 135

View Full Product Details