Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LETM1
Entrez Gene ID
3954
Gene Name
leucine zipper and EF-hand containing transmembrane protein 1
Gene Aliases
-
Chromosome Location
Chr.4: 1811479 - 1856247 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012318.2 NP_036450.1 6 - 7 1379 78
XM_006713884.1 XP_006713947.1 6 - 7 1087 78
XM_017008187.1 XP_016863676.1 5 - 6 1500 78
GenBank mRNA AF061025.1 - 6 - 7 1379 78
BC014500.1 - 6 - 7 1279 78
BC021208.1 - 6 - 7 1304 78

View Full Product Details