The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320226.1 | NP_001307155.1 | 2 - 3 | 297 | 98 |
NM_018032.4 | NP_060502.1 | 2 - 3 | 297 | 98 | |
NM_201412.2 | NP_958815.1 | 2 - 3 | 297 | 98 | |
XM_005255427.3 | XP_005255484.1 | 3 - 4 | 334 | 98 | |
XM_005255429.3 | XP_005255486.1 | 3 - 4 | 316 | 98 | |
XM_017023437.1 | XP_016878926.1 | 2 - 3 | 1360 | 98 | |
XM_017023438.1 | XP_016878927.1 | 2 - 3 | 1360 | 98 | |
XM_017023439.1 | XP_016878928.1 | 2 - 3 | 1360 | 98 | |
XM_017023440.1 | XP_016878929.1 | 2 - 3 | 1360 | 98 | |
GenBank mRNA | AJ404326.1 | - | 2 - 3 | 225 | 98 |
AK001093.1 | - | 2 - 3 | 225 | 98 | |
AK223084.1 | - | 2 - 3 | 228 | 98 | |
AY005111.1 | - | 2 - 3 | 265 | 98 | |
BC003194.1 | - | 2 - 3 | 242 | 98 | |
BC065198.1 | - | 2 - 3 | 1514 | 98 | |
BX647161.1 | - | 2 - 3 | 1295 | 98 | |
Z84716.2 | - | 1 - 2 | 648 | 98 |