The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005408.1 | NP_001005408.1 | 13 - 14 | 2345 | 97 |
NM_032442.2 | NP_115818.2 | 13 - 14 | 2345 | 97 | |
GenBank mRNA | AB058690.1 | - | 13 - 14 | 2345 | 97 |
AL136870.1 | - | 13 - 14 | 2201 | 97 | |
AL833570.1 | - | 13 - 14 | 1591 | 97 | |
BC007227.1 | - | 2 - 3 | 137 | 97 |