The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005408.1 | NP_001005408.1 | 18 - 19 | 3100 | 108 |
NM_032442.2 | NP_115818.2 | 18 - 19 | 3106 | 108 | |
GenBank mRNA | AB058690.1 | - | 18 - 19 | 3106 | 108 |
AL136870.1 | - | 18 - 19 | 2956 | 108 | |
AL833570.1 | - | 18 - 19 | 2352 | 108 | |
BC007227.1 | - | 7 - 8 | 892 | 108 |