The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000521.3 | NP_000512.1 | 2 - 3 | 553 | 68 |
NM_001292004.1 | NP_001278933.1 | 2 - 3 | 673 | 68 | |
GenBank mRNA | AF378118.1 | - | 2 - 3 | 508 | 68 |
AK122992.1 | - | 3 - 4 | 410 | 68 | |
AK130002.1 | - | 2 - 3 | 537 | 68 | |
AK130375.1 | - | 2 - 3 | 464 | 68 | |
AY643499.1 | - | 2 - 3 | 678 | 68 | |
BC017378.2 | - | 2 - 3 | 458 | 68 | |
BE907813.1 | - | 2 - 3 | 431 | 68 | |
BT009919.1 | - | 2 - 3 | 436 | 68 | |
FJ224336.1 | - | 2 - 3 | 678 | 68 | |
GQ472222.1 | - | 2 - 3 | 678 | 68 | |
M13519.1 | - | 2 - 3 | 484 | 68 | |
M19735.1 | - | 2 - 3 | 400 | 68 |