The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004891.3 | NP_004882.1 | 1 - 2 | 80 | 106 |
GenBank mRNA | AF047440.1 | - | 1 - 2 | 54 | 106 |
BC020834.1 | - | 1 - 2 | 67 | 106 | |
BI601959.1 | - | 1 - 2 | 83 | 106 |