Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GINS4
Entrez Gene ID
84296
Gene Name
GINS complex subunit 4
Gene Aliases
SLD5
Chromosome Location
Chr.8: 41529206 - 41545046 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032336.2 NP_115712.1 4 - 5 506 115
XM_005273659.3 XP_005273716.1 4 - 5 415 115
GenBank mRNA AK095334.1 - 4 - 5 463 115
AK313373.1 - 4 - 5 451 115
BC005995.1 - 4 - 5 485 115

View Full Product Details