The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291860.1 | NP_001278789.1 | 52 - 53 | 6808 | 69 |
NM_005529.6 | NP_005520.4 | 52 - 53 | 6805 | 69 | |
XM_011541318.2 | XP_011539620.1 | 57 - 58 | 7289 | 69 | |
XM_017001120.1 | XP_016856609.1 | 53 - 54 | 6935 | 69 | |
XM_017001121.1 | XP_016856610.1 | 52 - 53 | 6884 | 69 | |
XM_017001122.1 | XP_016856611.1 | 52 - 53 | 6881 | 69 | |
GenBank mRNA | AB209851.1 | - | 4 - 5 | 545 | 69 |
M85289.1 | - | 52 - 53 | 6805 | 69 | |
X62515.1 | - | 52 - 53 | 6771 | 69 |