The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291860.1 | NP_001278789.1 | 75 - 76 | 10444 | 82 |
NM_005529.6 | NP_005520.4 | 75 - 76 | 10441 | 82 | |
XM_011541318.2 | XP_011539620.1 | 80 - 81 | 10925 | 82 | |
XM_017001120.1 | XP_016856609.1 | 76 - 77 | 10571 | 82 | |
XM_017001121.1 | XP_016856610.1 | 75 - 76 | 10520 | 82 | |
XM_017001122.1 | XP_016856611.1 | 75 - 76 | 10517 | 82 | |
GenBank mRNA | AB209851.1 | - | 27 - 28 | 4181 | 82 |
M85289.1 | - | 75 - 76 | 10441 | 82 | |
X62515.1 | - | 75 - 76 | 10407 | 82 |