The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004654.3 | NP_004645.2 | 1 - 2 | 821 | 93 |
GenBank mRNA | AF000986.2 | - | 1 - 2 | 821 | 93 |