Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
USP9Y
Entrez Gene ID
8287
Gene Name
ubiquitin specific peptidase 9, Y-linked
Gene Aliases
DFFRY, SPGFY2
Chromosome Location
Chr.Y: 12701231 - 12860844 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004654.3 NP_004645.2 37 - 38 7160 94
XM_017030078.1 XP_016885567.1 36 - 37 6315 94
GenBank mRNA AF000986.2 - 37 - 38 7160 94
Y13618.1 - 35 - 36 6272 94

View Full Product Details