The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005257787.4 | XP_005257844.1 | 1 - 2 | 745 | 68 |
GenBank mRNA | BC017299.2 | - | 1 - 2 | 130 | 68 |