Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
E2F8
Entrez Gene ID
79733
Gene Name
E2F transcription factor 8
Gene Aliases
E2F-8
Chromosome Location
Chr.11: 19224063 - 19241655 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256372.1 NP_001243301.1 1 - 2 158 74
GenBank mRNA AB231783.1 - 1 - 2 100 74
AK292688.1 - 1 - 2 123 74

View Full Product Details