The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001258446.1 | NP_001245375.1 | 3 - 4 | 365 | 97 |
NM_001258447.1 | NP_001245376.1 | 2 - 3 | 224 | 97 | |
NM_052963.2 | NP_443195.1 | 2 - 3 | 273 | 97 | |
XM_005250781.3 | XP_005250838.1 | 2 - 3 | 261 | 97 | |
GenBank mRNA | AF349017.1 | - | 2 - 3 | 262 | 97 |
AK302372.1 | - | 2 - 3 | 224 | 97 | |
AK303237.1 | - | 2 - 3 | 234 | 97 | |
AK308475.1 | - | 2 - 3 | 245 | 97 | |
AK309005.1 | - | 2 - 3 | 382 | 97 | |
AK311647.1 | - | 2 - 3 | 245 | 97 | |
AK315913.1 | - | 2 - 3 | 235 | 97 | |
AK316380.1 | - | 3 - 4 | 312 | 97 | |
BC044646.1 | - | 2 - 3 | 248 | 97 | |
BC052285.1 | - | 2 - 3 | 248 | 97 | |
BC071914.1 | - | 2 - 3 | 246 | 97 | |
CB149603.1 | - | 2 - 3 | 263 | 97 | |
DC375680.1 | - | 2 - 3 | 273 | 97 | |
DC399840.1 | - | 2 - 3 | 224 | 97 |