The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127241.2 | NP_001120713.1 | 1 - 2 | 252 | 69 |
GenBank mRNA | AF354655.1 | - | 1 - 2 | 252 | 69 |