The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001171967.1 | NP_001165438.1 | 2 - 3 | 353 | 98 |
NM_024805.2 | NP_079081.2 | 2 - 3 | 353 | 98 | |
GenBank mRNA | AK024825.1 | - | 2 - 3 | 341 | 98 |
BC014195.2 | - | 2 - 3 | 215 | 98 | |
BC057772.1 | - | 2 - 3 | 221 | 98 |