The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000289.5 | NP_000280.1 | 1 - 2 | 180 | 61 |
GenBank mRNA | AK312284.1 | - | 1 - 2 | 62 | 61 |
BC021203.2 | - | 1 - 2 | 32 | 61 | |
DA525437.1 | - | 1 - 2 | 180 | 61 | |
DA795106.1 | - | 1 - 2 | 165 | 61 | |
M26066.1 | - | 1 - 2 | 71 | 61 | |
U24183.1 | - | 1 - 2 | 71 | 61 |