The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001267560.1 | NP_001254489.1 | 1 - 2 | 229 | 151 |
GenBank mRNA | AK091118.1 | - | 1 - 2 | 182 | 151 |
AL555606.3 | - | 1 - 2 | 228 | 151 |