The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001308152.1 | NP_001295081.1 | 10 - 11 | 1249 | 82 |
NM_001308153.1 | NP_001295082.1 | 12 - 13 | 1455 | 82 | |
NM_014629.3 | NP_055444.2 | 11 - 12 | 1363 | 82 | |
XM_005266041.3 | XP_005266098.1 | 11 - 12 | 1305 | 82 | |
XM_011534767.2 | XP_011533069.1 | 9 - 10 | 1108 | 82 | |
XM_011534768.1 | XP_011533070.1 | 10 - 11 | 1228 | 82 | |
XM_011534770.2 | XP_011533072.1 | 10 - 11 | 1228 | 82 | |
XM_017014003.1 | XP_016869492.1 | 11 - 12 | 1268 | 82 | |
GenBank mRNA | AB002292.2 | - | 11 - 12 | 1362 | 82 |
AF009205.1 | - | 2 - 3 | 197 | 82 | |
BC036809.1 | - | 3 - 4 | 913 | 82 | |
BC040474.1 | - | 8 - 9 | 933 | 82 | |
BC112926.1 | - | 10 - 11 | 1237 | 82 | |
CR749570.1 | - | 12 - 13 | 1416 | 82 |