Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TANK
Entrez Gene ID
10010
Gene Name
TRAF family member associated NFKB activator
Gene Aliases
I-TRAF, ITRAF, TRAF2
Chromosome Location
Chr.2: 161136955 - 161236176 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005246207.3 XP_005246264.2 1 - 2 92 91

View Full Product Details