The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021072.3 | NP_066550.2 | 3 - 4 | 1042 | 102 |
GenBank mRNA | AF064876.1 | - | 3 - 4 | 654 | 102 |
AF488549.1 | - | 3 - 4 | 1042 | 102 |