The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030779.3 | NP_110406.1 | 10 - 11 | 2312 | 84 |
XM_011525308.2 | XP_011523610.1 | 10 - 11 | 2312 | 84 | |
XM_011525309.2 | XP_011523611.1 | 10 - 11 | 2312 | 84 | |
XM_017025175.1 | XP_016880664.1 | 10 - 11 | 2312 | 84 | |
XM_017025176.1 | XP_016880665.1 | 10 - 11 | 2279 | 84 | |
XM_017025179.1 | XP_016880668.1 | 11 - 12 | 2153 | 84 | |
XM_017025180.1 | XP_016880669.1 | 10 - 11 | 2148 | 84 | |
GenBank mRNA | AF311913.1 | - | 10 - 11 | 2243 | 84 |