The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002772.2 | NP_002763.2 | 23 - 24 | 2797 | 93 |
XM_011529654.1 | XP_011527956.1 | 25 - 26 | 3071 | 93 | |
XM_011529655.1 | XP_011527957.1 | 26 - 27 | 2989 | 93 | |
XM_011529656.2 | XP_011527958.1 | 26 - 27 | 2996 | 93 | |
XM_011529657.1 | XP_011527959.1 | 24 - 25 | 3026 | 93 | |
XM_011529658.1 | XP_011527960.1 | 23 - 24 | 2990 | 93 | |
XM_011529659.1 | XP_011527961.1 | 24 - 25 | 2961 | 93 | |
GenBank mRNA | BC111749.1 | - | 23 - 24 | 2786 | 93 |
KF688184.1 | - | 3 - 4 | 414 | 93 | |
U09860.1 | - | 23 - 24 | 2806 | 93 |