The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002772.2 | NP_002763.2 | 2 - 3 | 301 | 154 |
XM_011529654.1 | XP_011527956.1 | 2 - 3 | 440 | 244 | |
XM_011529655.1 | XP_011527957.1 | 3 - 4 | 358 | 244 | |
XM_011529656.2 | XP_011527958.1 | 3 - 4 | 365 | 244 | |
XM_011529657.1 | XP_011527959.1 | 2 - 3 | 440 | 244 | |
XM_011529658.1 | XP_011527960.1 | 2 - 3 | 440 | 244 | |
XM_011529659.1 | XP_011527961.1 | 2 - 3 | 420 | 154 | |
GenBank mRNA | BC111749.1 | - | 2 - 3 | 290 | 154 |
DB242953.1 | - | 2 - 3 | 420 | 154 | |
U09860.1 | - | 2 - 3 | 310 | 154 |