The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002998.3 | NP_002989.2 | 4 - 5 | 1058 | 142 |
XM_011517212.2 | XP_011515514.1 | 5 - 6 | 590 | 142 | |
GenBank mRNA | AK074530.1 | - | 5 - 6 | 844 | 142 |
AK097839.1 | - | 5 - 6 | 859 | 142 | |
AK130131.1 | - | 4 - 5 | 899 | 142 | |
AK298643.1 | - | 4 - 5 | 758 | 142 | |
AK298939.1 | - | 4 - 5 | 600 | 142 | |
AK300067.1 | - | 4 - 5 | 796 | 142 | |
BC030133.2 | - | 4 - 5 | 1044 | 142 | |
BC049836.1 | - | 4 - 5 | 776 | 142 | |
J04621.1 | - | 4 - 5 | 1028 | 142 |