Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD44
Entrez Gene ID
960
Gene Name
CD44 molecule (Indian blood group)
Gene Aliases
CDW44, CSPG8, ECMR-III, HCELL, HUTCH-I, IN, LHR, MC56, MDU2, MDU3, MIC4, Pgp1
Chromosome Location
Chr.11: 35138870 - 35232402 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005253239.2 XP_005253296.1 7 - 8 1080 123
XM_005253240.2 XP_005253297.1 6 - 7 951 123
GenBank mRNA M83327.1 - 2 - 3 142 123

View Full Product Details