Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPAG16
Entrez Gene ID
79582
Gene Name
sperm associated antigen 16
Gene Aliases
PF20, WDR29
Chromosome Location
Chr.2: 213284369 - 214410501 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025436.2 NP_001020607.1 1 - 2 246 105
NM_024532.4 NP_078808.3 1 - 2 246 105
NR_047660.1 1 - 2 246 105
XM_006712746.2 XP_006712809.1 1 - 2 197 105
XM_011511815.1 XP_011510117.1 1 - 2 197 105
XM_011511816.2 XP_011510118.1 1 - 2 197 105
XM_011511817.1 XP_011510119.1 1 - 2 197 105
XM_011511818.1 XP_011510120.1 1 - 2 197 105
XM_011511820.1 XP_011510122.1 1 - 2 197 105
XM_011511824.1 XP_011510126.1 1 - 2 197 105
XM_011511827.1 XP_011510129.1 1 - 2 197 105
XM_011511828.1 XP_011510130.1 1 - 2 197 105
XM_011511829.1 XP_011510131.1 1 - 2 197 105
XM_011511831.1 XP_011510133.1 1 - 2 197 105
XM_011511832.1 XP_011510134.1 1 - 2 197 105
XM_011511837.2 XP_011510139.1 1 - 2 197 105
XM_017004896.1 XP_016860385.1 1 - 2 256 105
XM_017004899.1 XP_016860388.1 1 - 2 197 105
XM_017004900.1 XP_016860389.1 1 - 2 188 105
XM_017004901.1 XP_016860390.1 1 - 2 178 105
GenBank mRNA AF310672.1 - 1 - 2 183 105
AF426740.1 - 1 - 2 183 105
AF490390.1 - 1 - 2 176 105
AK026377.1 - 1 - 2 206 105
AK302351.1 - 1 - 2 188 105
AK310883.1 - 1 - 2 145 105
AL832962.1 - 1 - 2 135 105
BC009614.1 - 1 - 2 207 105
BC067756.1 - 1 - 2 233 105
BC100282.1 - 1 - 2 194 105
CR749477.1 - 1 - 2 171 123

View Full Product Details