Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPAG16
Entrez Gene ID
79582
Gene Name
sperm associated antigen 16
Gene Aliases
PF20, WDR29
Chromosome Location
Chr.2: 213284369 - 214410501 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025436.2 NP_001020607.1 2 - 3 291 86
NM_024532.4 NP_078808.3 2 - 3 291 86
NR_047660.1 2 - 3 291 86
XM_006712746.2 XP_006712809.1 2 - 3 242 86
XM_011511815.1 XP_011510117.1 2 - 3 242 86
XM_011511816.2 XP_011510118.1 2 - 3 242 86
XM_011511817.1 XP_011510119.1 2 - 3 242 86
XM_011511818.1 XP_011510120.1 2 - 3 242 86
XM_011511820.1 XP_011510122.1 2 - 3 242 86
XM_011511821.2 XP_011510123.1 2 - 3 1235 86
XM_011511824.1 XP_011510126.1 2 - 3 242 86
XM_011511827.1 XP_011510129.1 2 - 3 242 86
XM_011511828.1 XP_011510130.1 2 - 3 242 86
XM_011511829.1 XP_011510131.1 2 - 3 242 86
XM_011511831.1 XP_011510133.1 2 - 3 242 86
XM_011511832.1 XP_011510134.1 2 - 3 242 86
XM_011511837.2 XP_011510139.1 2 - 3 242 86
XM_017004896.1 XP_016860385.1 2 - 3 301 86
XM_017004899.1 XP_016860388.1 2 - 3 242 86
XM_017004900.1 XP_016860389.1 2 - 3 233 86
XM_017004901.1 XP_016860390.1 2 - 3 223 86
GenBank mRNA AF310672.1 - 2 - 3 228 86
AF426740.1 - 2 - 3 228 86
AF490390.1 - 2 - 3 221 86
AK026377.1 - 2 - 3 251 86
AK302351.1 - 2 - 3 233 86
AK310883.1 - 2 - 3 190 86
AL832962.1 - 2 - 3 180 86
BC009614.1 - 2 - 3 252 86
BC067756.1 - 2 - 3 278 86
BC100282.1 - 2 - 3 239 86

View Full Product Details