Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPAG16
Entrez Gene ID
79582
Gene Name
sperm associated antigen 16
Gene Aliases
PF20, WDR29
Chromosome Location
Chr.2: 213284369 - 214410501 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024532.4 NP_078808.3 6 - 7 752 144
NR_047659.1 8 - 9 927 144
NR_047660.1 5 - 6 633 144
XM_006712746.2 XP_006712809.1 6 - 7 703 144
XM_011511814.1 XP_011510116.1 5 - 6 800 144
XM_011511815.1 XP_011510117.1 6 - 7 703 144
XM_011511816.2 XP_011510118.1 6 - 7 703 144
XM_011511817.1 XP_011510119.1 6 - 7 703 144
XM_011511818.1 XP_011510120.1 6 - 7 703 144
XM_011511819.2 XP_011510121.1 5 - 6 1451 144
XM_011511820.1 XP_011510122.1 6 - 7 703 144
XM_011511821.2 XP_011510123.1 5 - 6 1577 144
XM_011511822.1 XP_011510124.1 4 - 5 433 144
XM_011511823.2 XP_011510125.1 4 - 5 1355 144
XM_011511824.1 XP_011510126.1 6 - 7 703 144
XM_011511827.1 XP_011510129.1 6 - 7 703 144
XM_011511828.1 XP_011510130.1 6 - 7 703 144
XM_011511829.1 XP_011510131.1 6 - 7 703 144
XM_011511831.1 XP_011510133.1 6 - 7 703 144
XM_011511832.1 XP_011510134.1 6 - 7 703 144
XM_011511835.2 XP_011510137.1 5 - 6 795 144
XM_011511837.2 XP_011510139.1 6 - 7 703 144
XM_017004896.1 XP_016860385.1 7 - 8 832 144
XM_017004897.1 XP_016860386.1 4 - 5 3449 144
XM_017004899.1 XP_016860388.1 6 - 7 703 144
XM_017004900.1 XP_016860389.1 6 - 7 694 144
GenBank mRNA AF310672.1 - 6 - 7 689 144
AF490390.1 - 6 - 7 682 144
AK095036.1 - 5 - 6 800 144
AK302351.1 - 5 - 6 575 144
AK310883.1 - 6 - 7 651 144
AL832962.1 - 6 - 7 641 144
BC025379.1 - 8 - 9 932 144
CR749477.1 - 4 - 5 438 144

View Full Product Details