Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UVRAG
Entrez Gene ID
7405
Gene Name
UV radiation resistance associated
Gene Aliases
DHTX, VPS38, p63
Chromosome Location
Chr.11: 75814852 - 76144238 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003369.3 NP_003360.2 6 - 7 832 79
XM_011545226.2 XP_011543528.1 6 - 7 1147 79
XM_011545227.2 XP_011543529.1 6 - 7 1147 79
XM_011545228.2 XP_011543530.1 6 - 7 1147 79
XM_017018224.1 XP_016873713.1 4 - 5 586 79
XM_017018225.1 XP_016873714.1 4 - 5 586 79
XM_017018226.1 XP_016873715.1 6 - 7 1147 79
GenBank mRNA AB012958.1 - 6 - 7 769 79
AK222772.1 - 6 - 7 723 79
AK291011.1 - 6 - 7 810 79
AK293782.1 - 6 - 7 573 79
BC064837.1 - 6 - 7 742 79

View Full Product Details