The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001283036.1 | NP_001269965.1 | 1 - 2 | 180 | 71 |
NM_001283037.1 | NP_001269966.1 | 1 - 2 | 180 | 71 | |
NM_016407.4 | NP_057491.2 | 1 - 2 | 180 | 71 | |
GenBank mRNA | AF117231.1 | - | 1 - 2 | 128 | 71 |
AF161513.1 | - | 1 - 2 | 137 | 71 | |
AF161518.1 | - | 1 - 2 | 106 | 71 | |
AF212244.1 | - | 1 - 2 | 117 | 71 | |
AK000481.1 | - | 1 - 2 | 110 | 71 | |
AK026118.1 | - | 1 - 2 | 108 | 71 | |
AK225135.1 | - | 1 - 2 | 98 | 71 | |
AK293662.1 | - | 1 - 2 | 108 | 71 | |
BC003359.1 | - | 1 - 2 | 107 | 71 | |
BG468509.1 | - | 1 - 2 | 137 | 71 | |
BP307634.1 | - | 1 - 2 | 174 | 71 | |
BU537939.1 | - | 1 - 2 | 90 | 71 | |
CR983692.1 | - | 1 - 2 | 91 | 71 |