The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002858.3 | NP_002849.1 | 15 - 16 | 1430 | 78 |
XM_006710802.2 | XP_006710865.2 | 16 - 17 | 1793 | 78 | |
XM_011541877.1 | XP_011540179.1 | 8 - 9 | 676 | 78 | |
XM_017001952.1 | XP_016857441.1 | 15 - 16 | 1971 | 78 | |
XM_017001953.1 | XP_016857442.1 | 15 - 16 | 1351 | 78 | |
XM_017001954.1 | XP_016857443.1 | 14 - 15 | 1411 | 78 | |
GenBank mRNA | AK296791.1 | - | 16 - 17 | 1466 | 78 |
AK302714.1 | - | 14 - 15 | 1355 | 78 | |
BC068509.1 | - | 11 - 12 | 1061 | 78 | |
BX648715.1 | - | 6 - 7 | 2178 | 78 | |
M81182.1 | - | 15 - 16 | 1328 | 78 | |
X58528.1 | - | 15 - 16 | 1352 | 78 |