The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002858.3 | NP_002849.1 | 9 - 10 | 930 | 139 |
XM_006710802.2 | XP_006710865.2 | 10 - 11 | 1293 | 139 | |
XM_017001952.1 | XP_016857441.1 | 9 - 10 | 1471 | 139 | |
XM_017001953.1 | XP_016857442.1 | 9 - 10 | 851 | 139 | |
XM_017001954.1 | XP_016857443.1 | 8 - 9 | 911 | 139 | |
GenBank mRNA | AK296791.1 | - | 10 - 11 | 966 | 139 |
AK302714.1 | - | 8 - 9 | 855 | 139 | |
AK308159.1 | - | 9 - 10 | 890 | 139 | |
M81182.1 | - | 9 - 10 | 828 | 139 | |
X58528.1 | - | 9 - 10 | 852 | 139 |