Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- UTP4
- Entrez Gene ID
-
84916
- Gene Name
- UTP4, small subunit processome component
- Gene Aliases
-
CIRH1A, CIRHIN, NAIC, TEX292
- Chromosome Location
-
Chr.16: 69132596 - 69169034 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK027445.1 |
- |
7 - 8 |
818 |
95 |
View Full Product Details