The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128429.2 | NP_001121901.1 | 15 - 16 | 2222 | 61 |
NM_001128430.1 | NP_001121902.1 | 15 - 16 | 2124 | 61 | |
NM_001254949.1 | NP_001241878.1 | 7 - 8 | 1044 | 61 | |
NM_020159.4 | NP_064544.2 | 15 - 16 | 2222 | 61 | |
NR_045644.1 | 16 - 17 | 2273 | 61 | ||
XM_017008463.1 | XP_016863952.1 | 15 - 16 | 2046 | 61 | |
XM_017008464.1 | XP_016863953.1 | 16 - 17 | 2223 | 61 | |
XM_017008465.1 | XP_016863954.1 | 16 - 17 | 2233 | 61 | |
GenBank mRNA | AB032948.2 | - | 14 - 15 | 2010 | 61 |
AK025302.1 | - | 10 - 11 | 1262 | 61 | |
AK027490.1 | - | 15 - 16 | 2120 | 61 | |
AK301668.1 | - | 7 - 8 | 1044 | 61 | |
AL359929.1 | - | 6 - 7 | 652 | 61 | |
AY008271.1 | - | 15 - 16 | 2025 | 61 | |
BC045534.1 | - | 15 - 16 | 2177 | 61 |