The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128429.2 | NP_001121901.1 | 19 - 20 | 2691 | 86 |
NM_001128430.1 | NP_001121902.1 | 19 - 20 | 2593 | 86 | |
NM_001254949.1 | NP_001241878.1 | 11 - 12 | 1507 | 86 | |
NM_020159.4 | NP_064544.2 | 19 - 20 | 2685 | 86 | |
NR_045644.1 | 20 - 21 | 2742 | 86 | ||
XM_017008463.1 | XP_016863952.1 | 19 - 20 | 2509 | 86 | |
XM_017008464.1 | XP_016863953.1 | 20 - 21 | 2692 | 86 | |
XM_017008465.1 | XP_016863954.1 | 20 - 21 | 2702 | 86 | |
GenBank mRNA | AB032948.2 | - | 18 - 19 | 2479 | 86 |
AK023990.1 | - | 4 - 5 | 474 | 86 | |
AK025302.1 | - | 14 - 15 | 1725 | 86 | |
AK301668.1 | - | 11 - 12 | 1507 | 86 | |
AL359929.1 | - | 10 - 11 | 1115 | 86 | |
AL512768.1 | - | 4 - 5 | 457 | 86 | |
AY008271.1 | - | 19 - 20 | 2488 | 86 | |
BC045534.1 | - | 19 - 20 | 2646 | 86 |