Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STXBP5
Entrez Gene ID
134957
Gene Name
syntaxin binding protein 5
Gene Aliases
LGL3, LLGL3, Nbla04300
Chromosome Location
Chr.6: 147204184 - 147390476 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127715.2 NP_001121187.1 22 - 23 2481 87
XM_017010267.1 XP_016865756.1 17 - 18 2552 87
GenBank mRNA AK090549.1 - 12 - 13 1191 87

View Full Product Details