Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STXBP5
Entrez Gene ID
134957
Gene Name
syntaxin binding protein 5
Gene Aliases
LGL3, LLGL3, Nbla04300
Chromosome Location
Chr.6: 147204184 - 147390476 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127715.2 NP_001121187.1 23 - 24 2721 79
NM_139244.4 NP_640337.3 21 - 22 2613 79
XM_006715339.2 XP_006715402.1 22 - 23 2847 79
XM_011535453.1 XP_011533755.1 22 - 23 2445 79
XM_017010267.1 XP_016865756.1 18 - 19 2792 79
GenBank mRNA AB073398.1 - 12 - 13 1452 79
AK055484.1 - 4 - 5 431 79
AK090549.1 - 13 - 14 1431 79
AK128121.1 - 22 - 23 2440 79
AL834152.1 - 24 - 25 2770 79
BC113382.1 - 21 - 22 2486 79
BC113408.1 - 21 - 22 2486 79

View Full Product Details