The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001856.3 | NP_001847.3 | 37 - 38 | 2909 | 110 |
XM_005270481.1 | XP_005270538.1 | 37 - 38 | 2904 | 110 | |
XM_011540722.1 | XP_011539024.1 | 37 - 38 | 2904 | 110 | |
XM_011540723.1 | XP_011539025.1 | 37 - 38 | 2904 | 110 | |
XM_011540724.1 | XP_011539026.1 | 36 - 37 | 2781 | 110 | |
XM_011540726.2 | XP_011539028.1 | 34 - 35 | 2420 | 110 | |
XM_011540727.2 | XP_011539029.1 | 27 - 28 | 1665 | 110 | |
XM_011540728.2 | XP_011539030.1 | 15 - 16 | 1147 | 110 | |
XM_011540729.1 | XP_011539031.1 | 37 - 38 | 2904 | 110 | |
XM_011540730.1 | XP_011539032.1 | 37 - 38 | 2904 | 110 | |
XM_017000338.1 | XP_016855827.1 | 37 - 38 | 2904 | 110 | |
XM_017000339.1 | XP_016855828.1 | 36 - 37 | 2868 | 110 | |
XM_017000340.1 | XP_016855829.1 | 14 - 15 | 1110 | 110 | |
XM_017000341.1 | XP_016855830.1 | 14 - 15 | 900 | 110 | |
GenBank mRNA | AB209571.1 | - | 33 - 34 | 2216 | 110 |
M92642.1 | - | 38 - 39 | 2694 | 110 | |
S57132.1 | - | 20 - 21 | 1290 | 110 |