Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COL16A1
Entrez Gene ID
1307
Gene Name
collagen type XVI alpha 1 chain
Gene Aliases
447AA, FP1572
Chromosome Location
Chr.1: 31652247 - 31704167 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001856.3 NP_001847.3 3 - 4 514 77
XM_005270481.1 XP_005270538.1 3 - 4 509 77
XM_011540722.1 XP_011539024.1 3 - 4 509 77
XM_011540723.1 XP_011539025.1 3 - 4 509 77
XM_011540724.1 XP_011539026.1 3 - 4 509 77
XM_011540729.1 XP_011539031.1 3 - 4 509 77
XM_011540730.1 XP_011539032.1 3 - 4 509 77
XM_017000338.1 XP_016855827.1 3 - 4 509 77
XM_017000339.1 XP_016855828.1 3 - 4 509 77
GenBank mRNA M92642.1 - 3 - 4 302 77

View Full Product Details