The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001856.3 | NP_001847.3 | 47 - 48 | 3470 | 101 |
XM_005270481.1 | XP_005270538.1 | 46 - 47 | 3417 | 101 | |
XM_011540724.1 | XP_011539026.1 | 46 - 47 | 3342 | 101 | |
XM_011540727.2 | XP_011539029.1 | 37 - 38 | 2226 | 101 | |
XM_011540728.2 | XP_011539030.1 | 25 - 26 | 1708 | 101 | |
XM_011540729.1 | XP_011539031.1 | 47 - 48 | 3465 | 101 | |
XM_011540730.1 | XP_011539032.1 | 46 - 47 | 3417 | 101 | |
XM_017000338.1 | XP_016855827.1 | 46 - 47 | 3432 | 101 | |
XM_017000339.1 | XP_016855828.1 | 46 - 47 | 3429 | 101 | |
XM_017000340.1 | XP_016855829.1 | 24 - 25 | 1671 | 101 | |
XM_017000341.1 | XP_016855830.1 | 24 - 25 | 1461 | 101 | |
GenBank mRNA | M92642.1 | - | 48 - 49 | 3255 | 101 |
S57132.1 | - | 30 - 31 | 1851 | 101 |