The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001856.3 | NP_001847.3 | 55 - 56 | 3856 | 57 |
XM_005270481.1 | XP_005270538.1 | 54 - 55 | 3803 | 57 | |
XM_011540722.1 | XP_011539024.1 | 54 - 55 | 3806 | 57 | |
XM_011540723.1 | XP_011539025.1 | 53 - 54 | 3758 | 57 | |
XM_011540724.1 | XP_011539026.1 | 54 - 55 | 3728 | 57 | |
XM_011540726.2 | XP_011539028.1 | 50 - 51 | 3274 | 57 | |
XM_011540727.2 | XP_011539029.1 | 45 - 46 | 2612 | 57 | |
XM_011540728.2 | XP_011539030.1 | 33 - 34 | 2094 | 57 | |
XM_017000338.1 | XP_016855827.1 | 54 - 55 | 3818 | 57 | |
XM_017000339.1 | XP_016855828.1 | 54 - 55 | 3815 | 57 | |
XM_017000340.1 | XP_016855829.1 | 32 - 33 | 2057 | 57 | |
XM_017000341.1 | XP_016855830.1 | 32 - 33 | 1847 | 57 | |
GenBank mRNA | AB209571.1 | - | 49 - 50 | 3070 | 57 |
S57132.1 | - | 38 - 39 | 2237 | 57 |