The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001856.3 | NP_001847.3 | 57 - 58 | 3992 | 97 |
XM_005270481.1 | XP_005270538.1 | 56 - 57 | 3939 | 97 | |
XM_011540722.1 | XP_011539024.1 | 56 - 57 | 3942 | 97 | |
XM_011540723.1 | XP_011539025.1 | 55 - 56 | 3894 | 97 | |
XM_011540724.1 | XP_011539026.1 | 56 - 57 | 3864 | 97 | |
XM_011540726.2 | XP_011539028.1 | 52 - 53 | 3410 | 97 | |
XM_011540727.2 | XP_011539029.1 | 47 - 48 | 2748 | 97 | |
XM_011540728.2 | XP_011539030.1 | 35 - 36 | 2230 | 97 | |
XM_017000338.1 | XP_016855827.1 | 56 - 57 | 3954 | 97 | |
XM_017000339.1 | XP_016855828.1 | 56 - 57 | 3951 | 97 | |
XM_017000340.1 | XP_016855829.1 | 34 - 35 | 2193 | 97 | |
XM_017000341.1 | XP_016855830.1 | 34 - 35 | 1983 | 97 | |
GenBank mRNA | AB209571.1 | - | 51 - 52 | 3206 | 97 |
AF370368.1 | - | 1 - 2 | 874 | 97 | |
M92642.1 | - | 58 - 59 | 3777 | 97 | |
S57132.1 | - | 40 - 41 | 2373 | 97 |