The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144073.1 | NP_001137545.1 | 7 - 8 | 1018 | 82 |
NM_012124.2 | NP_036256.2 | 8 - 9 | 1075 | 82 | |
XM_017017540.1 | XP_016873029.1 | 9 - 10 | 853 | 82 | |
XM_017017541.1 | XP_016873030.1 | 8 - 9 | 789 | 82 | |
GenBank mRNA | AF123249.1 | - | 7 - 8 | 713 | 82 |
AF217505.1 | - | 3 - 4 | 417 | 82 | |
AK225365.1 | - | 8 - 9 | 777 | 82 | |
AK225565.1 | - | 8 - 9 | 760 | 82 | |
AK226099.1 | - | 6 - 7 | 4077 | 82 | |
AK290231.1 | - | 8 - 9 | 1075 | 82 | |
AK312663.1 | - | 8 - 9 | 760 | 82 | |
BC017789.1 | - | 1 - 2 | 104 | 82 | |
BC072461.1 | - | 8 - 9 | 724 | 82 |